Product Details

SNP ID
rs11040908
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6517960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTATTCCTAATTGTTTTAAGGGA[A/G]AATCTGTGGATGGAAAATTTTCTGA
Phenotype
MIM: 615818
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNHD1 PubMed Links

Gene Details

Gene
DNHD1
Gene Name
dynein heavy chain domain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144666.2 Intron NP_653267.2
NM_173589.3 Intron NP_775860.3
Gene
RRP8
Gene Name
ribosomal RNA processing 8, methyltransferase, homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015324.3 Intron NP_056139.1
XM_011519955.2 Intron XP_011518257.1
XM_017017423.1 Intron XP_016872912.1

View Full Product Details