Product Details

SNP ID
rs12128292
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:224363891 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTTTTGTTGGGGTACCTGAAATG[A/G]CGGTGTTTTGGTAGGTGTTTTTTCG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNIH4 PubMed Links

Gene Details

Gene
CNIH4
Gene Name
cornichon family AMPA receptor auxiliary protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277197.1 Intron NP_001264126.1
NM_001277198.1 Intron NP_001264127.1
NM_001277199.1 Intron NP_001264128.1
NM_001277200.1 Intron NP_001264129.1
NM_014184.3 Intron NP_054903.1

View Full Product Details