Product Details
- SNP ID
-
rs3795441
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:226071232 on Build GRCh38
- Set Membership
-
JSNP
- Context Sequence [VIC/FAM]
- ATGTAAGCATTTGGTAAAATTGTCA[A/G]CATCTTGCCCAGTCATTTTTTTAAA
- Phenotype
-
MIM: 601128
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
H3F3A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77853027] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- H3F3A
- Gene Name
- H3 histone, family 3A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002107.4 |
|
Intron |
|
|
NP_002098.1 |
View Full Product Details