Product Details
- SNP ID
-
rs10926057
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
44
- Location
-
Chr.1:240009347 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGAAGCTGGGGAGGGGACACAGCA[G/T]TGATAAACTCAACCCAATTATGTGG
- Phenotype
-
MIM: 606373
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FMN2
PubMed Links
Gene Details
- Gene
- FMN2
- Gene Name
- formin 2
There are no transcripts associated with this gene.
- Gene
- RPS7P5
- Gene Name
- ribosomal protein S7 pseudogene 5
There are no transcripts associated with this gene.
View Full Product Details