Product Details

SNP ID
rs13027797
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:204548923 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAAGTAAGTCCATTATTTTCTGA[A/G]GCACTGAGTAATGTAGATGGGAGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARD3B PubMed Links

Gene Details

Gene
PARD3B
Gene Name
par-3 family cell polarity regulator beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302769.1 Intron NP_001289698.1
NM_057177.6 Intron NP_476518.4
NM_152526.5 Intron NP_689739.4
NM_205863.3 Intron NP_995585.2
XM_011510552.2 Intron XP_011508854.1
XM_011510553.2 Intron XP_011508855.1
XM_017003283.1 Intron XP_016858772.1
XM_017003284.1 Intron XP_016858773.1
XM_017003285.1 Intron XP_016858774.1
XM_017003286.1 Intron XP_016858775.1
XM_017003287.1 Intron XP_016858776.1
XM_017003288.1 Intron XP_016858777.1
XM_017003289.1 Intron XP_016858778.1
XM_017003290.1 Intron XP_016858779.1
XM_017003291.1 Intron XP_016858780.1
XM_017003292.1 Intron XP_016858781.1
XM_017003293.1 Intron XP_016858782.1
XM_017003294.1 Intron XP_016858783.1

View Full Product Details