Product Details

SNP ID
rs12117270
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:11107853 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTGCTCTAGCTTTTTCTTTCCTT[C/T]GCCCTTCCTGTATCTCTGCACTGGA
Phenotype
MIM: 605960 MIM: 601231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOSC10 PubMed Links

Gene Details

Gene
EXOSC10
Gene Name
exosome component 10
There are no transcripts associated with this gene.

Gene
LOC105376736
Gene Name
uncharacterized LOC105376736
There are no transcripts associated with this gene.

Gene
MTOR
Gene Name
mechanistic target of rapamycin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004958.3 Intron NP_004949.1
XM_005263438.2 Intron XP_005263495.1
XM_011541166.2 Intron XP_011539468.1
XM_017000900.1 Intron XP_016856389.1
XM_017000901.1 Intron XP_016856390.1
XM_017000902.1 Intron XP_016856391.1

View Full Product Details