Product Details
- SNP ID
-
hCV31724060
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:92219413 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTCAACTCAGCAGGAAGAACTTT[C/T]TAATTATTAAAGTTGTTGAAAAGTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
C1orf146
PubMed Links
Gene Details
- Gene
- C1orf146
- Gene Name
- chromosome 1 open reading frame 146
There are no transcripts associated with this gene.
View Full Product Details