Product Details

SNP ID
rs11230983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55773808 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGTTAGCAGCGATGGCTTATGACC[A/G]TTTTGTGGCAGTTTGTAAACCCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR5D13 PubMed Links

Gene Details

Gene
OR5D13
Gene Name
olfactory receptor family 5 subfamily D member 13 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001967.1 371 Missense Mutation CAT,CGT H124R NP_001001967.1

View Full Product Details