Product Details

SNP ID
rs4939473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60895937 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTATGGGCACATTTCATGGTGCC[C/G]CCCCGCCCAACAATTACAATAGCAA
Phenotype
MIM: 608330
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRPF19 PubMed Links

Gene Details

Gene
PRPF19
Gene Name
pre-mRNA processing factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014502.4 Intron NP_055317.1

View Full Product Details