Product Details

SNP ID
rs10736654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66078259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGTATGCGTCTTAGAAATTGTG[A/C]TGTGGGTAATTTGCTGTGTATGTTT
Phenotype
MIM: 607492 MIM: 605591
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PACS1 PubMed Links

Gene Details

Gene
PACS1
Gene Name
phosphofurin acidic cluster sorting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018026.3 Intron NP_060496.2
XM_011545162.1 Intron XP_011543464.1
XM_011545164.2 Intron XP_011543466.1
Gene
SF3B2
Gene Name
splicing factor 3b subunit 2
There are no transcripts associated with this gene.

View Full Product Details