Product Details

SNP ID
rs6591325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68000705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAGTGGTCACACACACTGGCAT[A/T]ACCCTTTGGGAAAGCAGCCTGGTAG
Phenotype
MIM: 600466 MIM: 608204
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ALDH3B1 PubMed Links

Gene Details

Gene
ALDH3B1
Gene Name
aldehyde dehydrogenase 3 family member B1
There are no transcripts associated with this gene.

Gene
UNC93B1
Gene Name
unc-93 homolog B1 (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030930.3 Intron NP_112192.2
XM_011545290.1 Intron XP_011543592.1
XM_011545291.2 Intron XP_011543593.1

View Full Product Details