Product Details

SNP ID
rs12354406
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:247858463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGTAGAACAACTGCCAGCACAG[A/G]GGAGGTCTAAGTCGTCTTTACTGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM58 PubMed Links

Gene Details

Gene
TRIM58
Gene Name
tripartite motif containing 58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015431.3 Intron NP_056246.3

View Full Product Details