Product Details
- SNP ID
-
rs10851420
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:45093718 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCAATTAGAGATGTGGGGAGCTAT[C/T]GGAGACAAGCACTATTGTACCTTTT
- Phenotype
-
MIM: 606759
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DUOX2
PubMed Links
Gene Details
- Gene
- DUOX2
- Gene Name
- dual oxidase 2
View Full Product Details