Product Details

SNP ID
rs6494134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59661432 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGTTTGCTTTCTCTTACTTCCTA[C/G]TTTTATCACTTTCCAATGCCTCTGC
Phenotype
MIM: 603292 MIM: 600519
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BNIP2 PubMed Links

Gene Details

Gene
BNIP2
Gene Name
BCL2 interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320674.1 4075 UTR 3 NP_001307603.1
NM_001320675.1 4075 UTR 3 NP_001307604.1
NM_004330.3 4075 UTR 3 NP_004321.2
Gene
GTF2A2
Gene Name
general transcription factor IIA subunit 2
There are no transcripts associated with this gene.

View Full Product Details