Product Details
- SNP ID
-
rs4776960
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:64174052 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTCTTCTGAATCATGATTTTCA[C/G]AGGTATTACTAACTTTTAAGTCAAC
- Phenotype
-
MIM: 606274
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CSNK1G1
PubMed Links
Gene Details
- Gene
- CSNK1G1
- Gene Name
- casein kinase 1 gamma 1
There are no transcripts associated with this gene.
View Full Product Details