Product Details

SNP ID
rs4238425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64378994 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGACCTTTATCAAGTTATATAAC[A/C]AGATATTAAATTTCCTTATCTTTAA
Phenotype
MIM: 610696 MIM: 604501
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0101 PubMed Links

Gene Details

Gene
KIAA0101
Gene Name
KIAA0101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029989.2 Intron NP_001025160.1
NM_014736.5 Intron NP_055551.1
Gene
TRIP4
Gene Name
thyroid hormone receptor interactor 4
There are no transcripts associated with this gene.

View Full Product Details