Product Details
- SNP ID
-
rs12595056
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:68297508 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATCATACACCTGTATCCAGAGTT[C/T]TGTTTTGTTTTTTTTTTTAGGTTTT
- Phenotype
-
MIM: 613539
MIM: 604789
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FEM1B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147618103,rs59986929] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FEM1B
- Gene Name
- fem-1 homolog B
There are no transcripts associated with this gene.
- Gene
- ITGA11
- Gene Name
- integrin subunit alpha 11
View Full Product Details