Product Details

SNP ID
rs10851784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68302792 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAGAGCCTGAGGGAGGCCTTGGC[A/G]TGGGCCTGGGTGTGTGTAGGGGTGT
Phenotype
MIM: 613539 MIM: 604789
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FEM1B PubMed Links

Gene Details

Gene
FEM1B
Gene Name
fem-1 homolog B
There are no transcripts associated with this gene.

Gene
ITGA11
Gene Name
integrin subunit alpha 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004439.1 3921 UTR 3 NP_001004439.1
XM_005254228.3 3921 UTR 3 XP_005254285.1
XM_011521363.2 3921 UTR 3 XP_011519665.1

View Full Product Details