Product Details

SNP ID
rs12632100
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194409996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGTAAGTTTTTAAGTCTTTAAA[A/G]AATGCACCTTAATCATGACCACAGA
Phenotype
MIM: 610232
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ATP13A3 PubMed Links

Gene Details

Gene
ATP13A3
Gene Name
ATPase 13A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024524.3 Intron NP_078800.3
XM_005269357.3 Intron XP_005269414.1
XM_011513120.2 Intron XP_011511422.1
XM_011513121.1 Intron XP_011511423.1
XM_011513122.1 Intron XP_011511424.1
XM_011513123.2 Intron XP_011511425.1
XM_011513124.2 Intron XP_011511426.1

View Full Product Details