Product Details

SNP ID
rs12489279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:171062327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTTAAATGCACCTGAGATTGCC[A/G]GTTGAGCTCTCTATATGTCTCCTGG
Phenotype
MIM: 610005
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNIK PubMed Links

Gene Details

Gene
TNIK
Gene Name
TRAF2 and NCK interacting kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161560.2 5895 UTR 3 NP_001155032.1
NM_001161561.2 5895 UTR 3 NP_001155033.1
NM_001161562.2 5895 UTR 3 NP_001155034.1
NM_001161563.2 5895 UTR 3 NP_001155035.1
NM_001161564.2 5895 UTR 3 NP_001155036.1
NM_001161565.2 5895 UTR 3 NP_001155037.1
NM_001161566.2 5895 UTR 3 NP_001155038.1
NM_015028.3 5895 UTR 3 NP_055843.1

View Full Product Details