Product Details
- SNP ID
-
rs13306057
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:129975823 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTAACCCAGACGCCGCGATGCCC[A/G]GCCCTTGGTTGCTGCTCGCTCTGGC
- Phenotype
-
MIM: 613879
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TRH
PubMed Links
Gene Details
- Gene
- TRH
- Gene Name
- thyrotropin releasing hormone
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007117.4 |
446 |
Missense Mutation |
AGC,GGC |
S3G |
NP_009048.1 |
View Full Product Details