Product Details

SNP ID
hCV31750181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86768246 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGATAATAATAAAGTAACGTTTTT[C/T]TAAGAATAAATATAAATATCTTAAA
Phenotype
MIM: 601299
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMPR1A PubMed Links

Gene Details

Gene
BMPR1A
Gene Name
bone morphogenetic protein receptor type 1A
There are no transcripts associated with this gene.

View Full Product Details