Product Details

SNP ID
rs12512664
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7460042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTAGGCGAGGGGCATTTAGGGT[A/G]ACTGAGCTGCTGCCGGGGCCTGGCG
Phenotype
MIM: 606284
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4274 PubMed Links

Gene Details

Gene
MIR4274
Gene Name
microRNA 4274
There are no transcripts associated with this gene.

Gene
SORCS2
Gene Name
sortilin related VPS10 domain containing receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020777.2 Intron NP_065828.2
XM_005247987.4 Intron XP_005248044.2
XM_011513514.2 Intron XP_011511816.1
XM_011513515.2 Intron XP_011511817.1
XM_011513516.2 Intron XP_011511818.1
XM_017008481.1 Intron XP_016863970.1

View Full Product Details