Product Details

SNP ID
rs10937784
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:7059208 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTTTGAAACAATTCTGGTTCTAA[C/G]CATTTCAGATGATAACCAACTTATA
Phenotype
MIM: 606173 MIM: 608790
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GRPEL1 PubMed Links

Gene Details

Gene
GRPEL1
Gene Name
GrpE like 1, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025196.3 2486 UTR 3 NP_079472.1
Gene
TADA2B
Gene Name
transcriptional adaptor 2B
There are no transcripts associated with this gene.

View Full Product Details