Product Details

SNP ID
rs11729435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4293947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTGCAGGAGGGAAGGAGACCTA[G/T]ACTCCACTACCCCATGGGAGAATCA
Phenotype
MIM: 616238
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LYAR PubMed Links

Gene Details

Gene
LYAR
Gene Name
Ly1 antibody reactive
There are no transcripts associated with this gene.

Gene
ZBTB49
Gene Name
zinc finger and BTB domain containing 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145291.3 Intron NP_660334.3
XM_005247951.4 Intron XP_005248008.1
XM_006713864.3 Intron XP_006713927.1
XM_011513416.1 Intron XP_011511718.1
XM_017007835.1 Intron XP_016863324.1
XM_017007836.1 Intron XP_016863325.1

View Full Product Details