Product Details

SNP ID
rs13131946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:711223 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCCTCCCACCATGCCGCACTCT[C/T]TAGACTACAGCGGCTCTGTGGCAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCGF3 PubMed Links

Gene Details

Gene
PCGF3
Gene Name
polycomb group ring finger 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317836.1 Intron NP_001304765.1
NM_006315.5 Intron NP_006306.2

View Full Product Details