Product Details

SNP ID
hCV31759145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56451763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGCCGCAGGACGCCGTTCAGT[C/T]ACGGTAACCGGGTGCTCTCAGGGAG
Phenotype
MIM: 604978 MIM: 615857
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUDT21 PubMed Links

Gene Details

Gene
NUDT21
Gene Name
nudix hydrolase 21
There are no transcripts associated with this gene.

Gene
OGFOD1
Gene Name
2-oxoglutarate and iron dependent oxygenase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details