Product Details

SNP ID
rs11568314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66936658 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCTTCTCCAGGCCCTTATCAGA[A/T]GCTGGGGGAGTGTTTCGGCCTTTGG
Phenotype
MIM: 605278 MIM: 614778
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CES2 PubMed Links

Gene Details

Gene
CES2
Gene Name
carboxylesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003869.5 Intron NP_003860.2
NM_198061.2 Intron NP_932327.1
XM_011523421.2 Intron XP_011521723.1
XM_017023818.1 Intron XP_016879307.1
Gene
FAM96B
Gene Name
family with sequence similarity 96 member B
There are no transcripts associated with this gene.

View Full Product Details