Product Details

SNP ID
rs12535098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:128302357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGGATCGAGGGTATGTGACTGC[C/T]TGAGCAGCTACTTCAATCCTAATGT
Phenotype
MIM: 612074
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGC27345 PubMed Links

Gene Details

Gene
MGC27345
Gene Name
uncharacterized protein MGC27345
There are no transcripts associated with this gene.

Gene
RBM28
Gene Name
RNA binding motif protein 28
There are no transcripts associated with this gene.

View Full Product Details