Product Details

SNP ID
rs13339753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:9333181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGACAGGGTTTCACCGTGTGTTGC[C/T]TAGGCTGGGCTCAAGCAATCTACCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF559 PubMed Links

Gene Details

Gene
ZNF559
Gene Name
zinc finger protein 559
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202406.1 Intron NP_001189335.1
NM_001202407.1 Intron NP_001189336.1
NM_001202408.1 Intron NP_001189337.1
NM_001202409.1 Intron NP_001189338.1
NM_001202410.1 Intron NP_001189339.1
NM_001202411.1 Intron NP_001189340.1
NM_001202412.1 Intron NP_001189341.1
NM_032497.2 Intron NP_115886.1
Gene
ZNF559-ZNF177
Gene Name
ZNF559-ZNF177 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172650.2 Intron NP_001166121.1
NM_001202425.1 Intron NP_001189354.1

View Full Product Details