Product Details

SNP ID
rs11085733
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10426323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGTGGCTGAGAATGCTGTAGAC[C/T]AAGTTTGTCCAACCCGTGGCCCAGG
Phenotype
MIM: 600126
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PDE4A PubMed Links

Gene Details

Gene
PDE4A
Gene Name
phosphodiesterase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111307.1 Intron NP_001104777.1
NM_001111308.1 Intron NP_001104778.1
NM_001111309.1 Intron NP_001104779.1
NM_001243121.1 Intron NP_001230050.1
NM_006202.2 Intron NP_006193.1
XM_005259933.2 Intron XP_005259990.1
XM_011528054.1 Intron XP_011526356.1
XM_011528055.2 Intron XP_011526357.1
XM_017026865.1 Intron XP_016882354.1

View Full Product Details