Product Details

SNP ID
rs11085996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:15836302 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATATGTGAAAAGGTGACAGAAAAGA[A/G]ATCTTCCTAAAAGAGTCAGCCACTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UCA1 PubMed Links

Gene Details

Gene
UCA1
Gene Name
urothelial cancer associated 1 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details