Product Details

SNP ID
rs3802341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130597785 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAAACAGTGCTCAACAGAAGACA[C/T]GATTAATTTTATCGAGCTGTAATAC
Phenotype
MIM: 603536
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUBP3 PubMed Links

Gene Details

Gene
FUBP3
Gene Name
far upstream element binding protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003934.1 Intron NP_003925.1
XM_005272232.1 Intron XP_005272289.1
XM_006717313.2 Intron XP_006717376.1
XM_011519172.2 Intron XP_011517474.1
XM_011519173.2 Intron XP_011517475.1
XM_011519174.2 Intron XP_011517476.1
XM_017015269.1 Intron XP_016870758.1
XM_017015270.1 Intron XP_016870759.1
XM_017015271.1 Intron XP_016870760.1
XM_017015272.1 Intron XP_016870761.1
XM_017015273.1 Intron XP_016870762.1

View Full Product Details