Product Details
- SNP ID
-
rs11244112
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:130724970 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCACAGGTAGACTTCGGAAGGTG[A/T]CAGGCCTTAGAGCCACAGTGGCCGC
- Phenotype
-
MIM: 189980
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ABL1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80005896] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ABL1
- Gene Name
- ABL proto-oncogene 1, non-receptor tyrosine kinase
View Full Product Details