Product Details

SNP ID
rs7298857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:31657192 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTACTTGCACCTGCAAGAGGGACC[A/G]ATTATTAGAGAACTGGCTTTGTAAA
Phenotype
MIM: 615256
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ETFBKMT PubMed Links

Gene Details

Gene
ETFBKMT
Gene Name
electron transfer flavoprotein beta subunit lysine methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135863.1 Intron NP_001129335.1
NM_001135864.1 Intron NP_001129336.1
NM_173802.3 Intron NP_776163.1

View Full Product Details