Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001286521.1 | 3242 | UTR 3 | NP_001273450.1 | ||
NM_001286522.1 | 3242 | UTR 3 | NP_001273451.1 | ||
NM_020367.5 | 3242 | UTR 3 | NP_065100.2 | ||
XM_005253711.4 | 3242 | Intron | XP_005253768.1 | ||
XM_011520970.1 | 3242 | Intron | XP_011519272.1 | ||
XM_011520971.2 | 3242 | Intron | XP_011519273.1 | ||
XM_011520973.2 | 3242 | Intron | XP_011519275.1 | ||
XM_017019670.1 | 3242 | Intron | XP_016875159.1 | ||
XM_017019671.1 | 3242 | Intron | XP_016875160.1 |