Product Details

SNP ID
rs10922241
Assay Type
Functionally tested
NCBI dbSNP Submissions
53
Location
Chr.1:197508107 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTGGATTTCTGAAGCCGTGATGT[C/T]GATAAAGCAAATGGACTAAAAACCA
Phenotype
MIM: 613292
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DENND1B PubMed Links

Gene Details

Gene
DENND1B
Gene Name
DENN domain containing 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195215.1 4493 UTR 3 NP_001182144.1
NM_001195216.1 4493 Intron NP_001182145.1
NM_001300858.1 4493 Intron NP_001287787.1
NM_144977.4 4493 Intron NP_659414.2
XM_005244931.2 4493 UTR 3 XP_005244988.1
XM_006711192.2 4493 UTR 3 XP_006711255.1
XM_006711193.3 4493 UTR 3 XP_006711256.1
XM_006711194.3 4493 UTR 3 XP_006711257.1
XM_011509246.2 4493 UTR 3 XP_011507548.1
XM_011509247.1 4493 UTR 3 XP_011507549.1
XM_011509248.2 4493 UTR 3 XP_011507550.1
XM_011509249.2 4493 UTR 3 XP_011507551.1
XM_011509251.2 4493 Intron XP_011507553.1
XM_017000470.1 4493 UTR 3 XP_016855959.1
XM_017000471.1 4493 UTR 3 XP_016855960.1
XM_017000472.1 4493 UTR 3 XP_016855961.1

View Full Product Details