Product Details

SNP ID
rs13261032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:119733117 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTATACCAGGGAGGGATCTTGGAA[A/C]TAATCCCCTCTGCGTATACCAAAGG
Phenotype
MIM: 604912
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TAF2 PubMed Links

Gene Details

Gene
TAF2
Gene Name
TATA-box binding protein associated factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003184.3 Intron NP_003175.1
XM_006716621.3 Intron XP_006716684.1
XM_011517259.2 Intron XP_011515561.1
XM_017013779.1 Intron XP_016869268.1
XM_017013780.1 Intron XP_016869269.1
XM_017013781.1 Intron XP_016869270.1
XM_017013782.1 Intron XP_016869271.1
XM_017013783.1 Intron XP_016869272.1
XM_017013784.1 Intron XP_016869273.1
XM_017013785.1 Intron XP_016869274.1
XM_017013786.1 Intron XP_016869275.1
XM_017013787.1 Intron XP_016869276.1
XM_017013788.1 Intron XP_016869277.1
XM_017013789.1 Intron XP_016869278.1
XM_017013790.1 Intron XP_016869279.1
XM_017013791.1 Intron XP_016869280.1

View Full Product Details