Product Details

SNP ID
rs6857207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38977937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTCTTCTTTTTCTTCTTTTAGA[A/T]AAAGATTTCTATCGCTTTTCAAAGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM156 PubMed Links

Gene Details

Gene
TMEM156
Gene Name
transmembrane protein 156
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303228.1 4076 Intron NP_001290157.1
NM_024943.2 4076 Intron NP_079219.1
XM_005262670.3 4076 Intron XP_005262727.1
XM_011513749.2 4076 Intron XP_011512051.1
XM_011513750.2 4076 Intron XP_011512052.1
XM_011513751.2 4076 UTR 3 XP_011512053.1
XM_011513752.2 4076 Intron XP_011512054.1
XM_011513753.2 4076 Intron XP_011512055.1
XM_011513754.2 4076 Intron XP_011512056.1
XM_017008629.1 4076 Intron XP_016864118.1

View Full Product Details