Product Details

SNP ID
rs13435456
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:38977987 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCTTTATATCCATGACATCTGGT[A/G]TCTAACCTCAACTCATCTTTCCCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM156 PubMed Links

Gene Details

Gene
TMEM156
Gene Name
transmembrane protein 156
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303228.1 4026 Intron NP_001290157.1
NM_024943.2 4026 Intron NP_079219.1
XM_005262670.3 4026 Intron XP_005262727.1
XM_011513749.2 4026 Intron XP_011512051.1
XM_011513750.2 4026 Intron XP_011512052.1
XM_011513751.2 4026 UTR 3 XP_011512053.1
XM_011513752.2 4026 Intron XP_011512054.1
XM_011513753.2 4026 Intron XP_011512055.1
XM_011513754.2 4026 Intron XP_011512056.1
XM_017008629.1 4026 Intron XP_016864118.1

View Full Product Details