Product Details

SNP ID
rs10863961
Assay Type
Functionally Tested
NCBI dbSNP Submissions
37
Location
Chr.1:212294505 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCCCTACCATTCTTCTTTAAATT[A/G]GTCAGTGGCAAAATATTTTGTTTGC
Phenotype
MIM: 601643
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
LOC101929541 PubMed Links
Additional Information
For this assay, SNP(s) [rs145906272] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LOC101929541
Gene Name
uncharacterized LOC101929541
There are no transcripts associated with this gene.

Gene
PPP2R5A
Gene Name
protein phosphatase 2 regulatory subunit B'alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199756.1 Intron NP_001186685.1
NM_006243.3 Intron NP_006234.1

View Full Product Details