Product Details

SNP ID
rs10737472
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:23382426 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCGTAATACTAGTAGATGACATA[A/C]CCAAAATAGTAAGAATAGTGGTTTT
Phenotype
MIM: 604128
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TCEA3 PubMed Links

Gene Details

Gene
TCEA3
Gene Name
transcription elongation factor A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003196.2 Intron NP_003187.1
XM_006710864.1 Intron XP_006710927.1
XM_011542053.2 Intron XP_011540355.1
XM_017002200.1 Intron XP_016857689.1
XM_017002201.1 Intron XP_016857690.1
XM_017002202.1 Intron XP_016857691.1
XM_017002203.1 Intron XP_016857692.1
XM_017002204.1 Intron XP_016857693.1

View Full Product Details