Product Details

SNP ID
rs12760004
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:23638319 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACAGCGAATGATAGTGACAGCAAC[C/T]GTGCTAAGAAGAGCTGCCACTGTGG
Phenotype
MIM: 607305
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MDS2 PubMed Links

Gene Details

Gene
MDS2
Gene Name
myelodysplastic syndrome 2 translocation associated
There are no transcripts associated with this gene.

View Full Product Details