Product Details

SNP ID
rs10917413
Assay Type
Functionally tested
NCBI dbSNP Submissions
49
Location
Chr.1:23691787 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGCCTCCTGGCCCATAAGGCCCT[C/G]GGCCGGAAGCTCCGCTTTCTCTTCC
Phenotype
MIM: 604175
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPL11 PubMed Links

Gene Details

Gene
RPL11
Gene Name
ribosomal protein L11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000975.3 Intron NP_000966.2
NM_001199802.1 Intron NP_001186731.1

View Full Product Details