Product Details

SNP ID
rs12622875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:124029560 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGATTTCCACACAAGTTCAAAAT[C/T]ATGAAAAAATTAGTTTTATTACGAA
Phenotype
MIM: 610519
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNTNAP5 PubMed Links

Gene Details

Gene
CNTNAP5
Gene Name
contactin associated protein like 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130773.3 Intron NP_570129.1
XM_006712258.1 Intron XP_006712321.1
XM_017003316.1 Intron XP_016858805.1

View Full Product Details