Product Details
- SNP ID
-
rs12304307
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:10453116 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGATTTTATTGTATATTAATTATAC[C/G]TCCTTTGTATCATATTGAGCACTTG
- Phenotype
-
MIM: 161555
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KLRC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114439078] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KLRC1
- Gene Name
- killer cell lectin like receptor C1
View Full Product Details