Product Details

SNP ID
rs3136390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:54181934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGGAGTAGTGTCAAAACTGCCA[C/T]GGCAGGTTGGAAGACAGTTCAACAG
Phenotype
MIM: 607753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMUG1 PubMed Links

Gene Details

Gene
SMUG1
Gene Name
single-strand-selective monofunctional uracil-DNA glycosylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243787.1 1076 UTR 3 NP_001230716.1
NM_001243788.1 1076 UTR 3 NP_001230717.1
NM_001243789.1 1076 Intron NP_001230718.1
NM_001243790.1 1076 Intron NP_001230719.1
NM_001243791.1 1076 Intron NP_001230720.1
NM_014311.2 1076 UTR 3 NP_055126.1
XM_006719319.3 1076 UTR 3 XP_006719382.1
XM_006719320.3 1076 UTR 3 XP_006719383.1
XM_006719321.3 1076 UTR 3 XP_006719384.1
XM_006719322.3 1076 Intron XP_006719385.1
XM_011538109.2 1076 UTR 3 XP_011536411.1
XM_011538110.2 1076 UTR 3 XP_011536412.1
XM_011538111.2 1076 UTR 3 XP_011536413.1
XM_011538112.2 1076 UTR 3 XP_011536414.1
XM_011538113.2 1076 UTR 3 XP_011536415.1
XM_011538114.2 1076 UTR 3 XP_011536416.1
XM_011538115.2 1076 UTR 3 XP_011536417.1
XM_011538116.2 1076 UTR 3 XP_011536418.1
XM_011538117.2 1076 UTR 3 XP_011536419.1
XM_011538118.2 1076 UTR 3 XP_011536420.1
XM_011538119.2 1076 UTR 3 XP_011536421.1
XM_011538120.2 1076 UTR 3 XP_011536422.1
XM_011538121.2 1076 UTR 3 XP_011536423.1
XM_011538122.2 1076 Intron XP_011536424.1
XM_011538123.2 1076 Intron XP_011536425.1
XM_017019113.1 1076 UTR 3 XP_016874602.1
XM_017019114.1 1076 UTR 3 XP_016874603.1
XM_017019115.1 1076 UTR 3 XP_016874604.1
XM_017019116.1 1076 UTR 3 XP_016874605.1
XM_017019117.1 1076 Intron XP_016874606.1
XM_017019118.1 1076 Intron XP_016874607.1
XM_017019119.1 1076 Intron XP_016874608.1
XM_017019120.1 1076 Intron XP_016874609.1

View Full Product Details