Product Details
- SNP ID
-
rs12309146
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:63789644 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCACAATAAAATGAGGTGTGCTGG[A/T]ATTATGGCCAAAGAGTAATACCGTT
- Phenotype
-
MIM: 605862
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77476543] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM5
- Gene Name
- transmembrane protein 5
View Full Product Details