Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001031701.2 | Intron | NP_001026871.1 | |||
XM_011538476.2 | Intron | XP_011536778.1 | |||
XM_017019454.1 | Intron | XP_016874943.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_017564.9 | Intron | NP_060034.9 | |||
XM_011538537.2 | Intron | XP_011536839.1 | |||
XM_011538538.2 | Intron | XP_011536840.1 | |||
XM_011538539.2 | Intron | XP_011536841.1 | |||
XM_011538541.2 | Intron | XP_011536843.1 | |||
XM_011538542.2 | Intron | XP_011536844.1 | |||
XM_017019585.1 | Intron | XP_016875074.1 |