Product Details
- SNP ID
-
rs12580300
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:111411467 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTAAAAGGAAAATTTCAGCCTCCC[A/G]AATTATACTATTATCCAGGGTGATT
- Phenotype
-
MIM: 605093
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SH2B3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114598006] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SH2B3
- Gene Name
- SH2B adaptor protein 3
View Full Product Details